SY3H3D67F5D5 Synthonix Corporation

2-Iododibenzo[b,d]furan

  • MDL number No MDL found
  • Cas Number : 5408-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/56403.html

Safety Information

MSDS https://synthonix.com/56403.html
Availability details for
Quantity
  • Availability
  • Add to Cart