SY3H3D67F5D6 Synthonix Corporation

3-Iodo-9H-carbazole

  • MDL number No MDL found
  • Cas Number : 16807-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56442.html

Safety Information

MSDS https://synthonix.com/56442.html
Availability details for
Quantity
  • Availability
  • Add to Cart