SY3H3D67F5D8 Synthonix Corporation

3-Iodo-biphenyl

  • MDL number No MDL found
  • Cas Number : 20442-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/56478.html

Safety Information

MSDS https://synthonix.com/56478.html
Availability details for
Quantity
  • Availability
  • Add to Cart