SY3H3D67F5DB Synthonix Corporation

4-(Indolin-5-yl)morpholine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1646152-53-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/56608.html

Safety Information

MSDS https://synthonix.com/56608.html
Availability details for
Quantity
  • Availability
  • Add to Cart