SY3H3D67F5DC Synthonix Corporation

5-(1H-Indol-4-yl)-1H-pyrrole-2-carboxylic acid

  • MDL number No MDL found
  • Cas Number : 2111055-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/56655.html

Safety Information

MSDS https://synthonix.com/56655.html
Availability details for
Quantity
  • Availability
  • Add to Cart