SY3H3D67F5E2 Synthonix Corporation

2-Iodo-9,9-dimethyl-9H-fluorene

  • Cas Number : 144981-85-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58172.html

Safety Information

MSDS https://synthonix.com/58172.html
Availability details for
Quantity
  • Availability
  • Add to Cart