SY3H3D67F5E3 Synthonix Corporation

4-Iodo-4'-propylbiphenyl

  • Cas Number : 782477-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/58250.html

Safety Information

MSDS https://synthonix.com/58250.html
Availability details for
Quantity
  • Availability
  • Add to Cart