SY3H3D67F5E4 Synthonix Corporation

2-(2-(1H-Indol-3-yl)acetamido)pentanedioic acid

  • MDL number No MDL found
  • Cas Number : 192430-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58272.html

Safety Information

MSDS https://synthonix.com/58272.html
Availability details for
Quantity
  • Availability
  • Add to Cart