SY3H3D67F5E6 Synthonix Corporation

(1-Tosyl-1H-indol-4-yl)methanamine

  • MDL number No MDL found
  • Cas Number : 1145678-74-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58670.html

Safety Information

MSDS https://synthonix.com/58670.html
Availability details for
Quantity
  • Availability
  • Add to Cart