SY3H3D67F5E7 Synthonix Corporation

1-(2-Iodoethyl)-4-octylbenzene

  • Cas Number : 162358-07-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/58834.html

Safety Information

MSDS https://synthonix.com/58834.html
Availability details for
Quantity
  • Availability
  • Add to Cart