SY3H3D67F5E8 Synthonix Corporation

3-Iodo-N-phenylcarbazole

  • MDL number No MDL found
  • Cas Number : 502161-03-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/59147.html

Safety Information

MSDS https://synthonix.com/59147.html
Availability details for
Quantity
  • Availability
  • Add to Cart