SY3H3D67F5E9 Synthonix Corporation

4-Iodo-N,N-diphenylaniline

  • MDL number No MDL found
  • Cas Number : 38257-52-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/59168.html

Safety Information

MSDS https://synthonix.com/59168.html
Availability details for
Quantity
  • Availability
  • Add to Cart