SY3H3D67F5EA Synthonix Corporation

N-(5-Iodo-4-oxo-7-tosyl-4,7-dihydro-3H-pyrrolo[2,3-d]pyrimidin-2-yl)pivalamide

  • MDL number No MDL found
  • Cas Number : 2097938-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/59215.html

Safety Information

MSDS https://synthonix.com/59215.html
Availability details for
Quantity
  • Availability
  • Add to Cart