SY3H3D67F5ED Synthonix Corporation

(E)-6-Iodo-3-(2-(pyridin-2-yl)vinyl)-1-(tetrahydro-2H-pyran-2-yl)-1H-indazole

  • MDL number No MDL found
  • Cas Number : 886230-77-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/59393.html

Safety Information

MSDS https://synthonix.com/59393.html
Availability details for
Quantity
  • Availability
  • Add to Cart