SY3H3D67F5F2 Synthonix Corporation

Lithium [1,2,4]triazolo[4,3-a]pyrazine-3-carboxylate

  • Cas Number : 1799434-66-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/41882.html

Safety Information

MSDS https://synthonix.com/41882.html
Availability details for
Quantity
  • Availability
  • Add to Cart