SY3H3D67F607 Synthonix Corporation

Methyl 3-oxohexanoate

  • Cas Number : 30414-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39290.html

Safety Information

MSDS https://synthonix.com/39290.html
Availability details for
Quantity
  • Availability
  • Add to Cart