SY3H3D67F611 Synthonix Corporation

N-Methyl-2-fluoroaniline

  • MDL number No MDL found
  • Cas Number : 1978-38-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39414.html

Safety Information

MSDS https://synthonix.com/39414.html
Availability details for
Quantity
  • Availability
  • Add to Cart