SY3H3D67F618 Synthonix Corporation

Methyl 4-chloro-3-nitrobenzate

  • Cas Number : 14719-83-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39468.html

Safety Information

MSDS https://synthonix.com/39468.html
Availability details for
Quantity
  • Availability
  • Add to Cart