SY3H3D67F61F Synthonix Corporation

4-(Methylsulfonyl)-2-nitrobenzoic acid

  • MDL number No MDL found
  • Cas Number : 110964-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39485.html

Safety Information

MSDS https://synthonix.com/39485.html
Availability details for
Quantity
  • Availability
  • Add to Cart