SY3H3D67F634 Synthonix Corporation

2-Methoxycinnamic Acid

  • MDL number MFCD00064238
  • Cas Number : 6099-03-2
  • Molecular Weight : 178.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerator
Physical Form Powder with lumps Colour: white - pale-yellow
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39590.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39590.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart