SY3H3D67F639 Synthonix Corporation

Methyl 3-phenylpropionate

  • MDL number No MDL found
  • Cas Number : 103-25-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39604.html

Safety Information

MSDS https://synthonix.com/39604.html
Availability details for
Quantity
  • Availability
  • Add to Cart