SY3H3D67F63A Synthonix Corporation

2-Methyl-2-phenylpropanoic acid

  • MDL number No MDL found
  • Cas Number : 826-55-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39605.html

Safety Information

MSDS https://synthonix.com/39605.html
Availability details for
Quantity
  • Availability
  • Add to Cart