SY3H3D67F63B Synthonix Corporation

Methyl non-2-enoate

  • MDL number MFCD00027316
  • Cas Number : 111-79-5
  • Molecular Weight : 170.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerator
Physical Form liquid Colour: colourless
Shipping Temperature Room Temperature
Country of Origin US
Product Link https://synthonix.com/39622.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39622.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart