SY3H3D67F63E Synthonix Corporation

5-(4-Methyl-1,3-dioxolan-2-yl)benzo[d][1,3]dioxole

  • Cas Number : 61683-99-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/39651.html

Safety Information

MSDS https://synthonix.com/39651.html
Availability details for
Quantity
  • Availability
  • Add to Cart