SY3H3D67F642 Synthonix Corporation

5-Methyl-2-phenylhex-2-enal

  • MDL number No MDL found
  • Cas Number : 21834-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/39706.html

Safety Information

MSDS https://synthonix.com/39706.html
Availability details for
Quantity
  • Availability
  • Add to Cart