SY3H3D67F647 Synthonix Corporation

N,N'-(Methylenebis(4,1-phenylene))bis(2,2-dimethylhydrazinecarboxamide)

  • MDL number No MDL found
  • Cas Number : 85095-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/39751.html

Safety Information

MSDS https://synthonix.com/39751.html
Availability details for
Quantity
  • Availability
  • Add to Cart