SY3H3D67F659 Synthonix Corporation

2-Methoxyethyl methanesulfonate

  • MDL number MFCD02656401
  • Cas Number : 16427-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Clear liquid / Colour: Colorless - Pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/39960.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/39960.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart