SY3H3D67F65F Synthonix Corporation

1-Methyl-4-nitroimidazole

  • Cas Number : 3034-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40134.html

Safety Information

MSDS https://synthonix.com/40134.html
Availability details for
Quantity
  • Availability
  • Add to Cart