SY3H3D67F664 Synthonix Corporation

Methyl 2H-1,2,3-triazole-4-carboxylate

  • Cas Number : 877309-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40141.html

Safety Information

MSDS https://synthonix.com/40141.html
Availability details for
Quantity
  • Availability
  • Add to Cart