SY3H3D67F666 Synthonix Corporation

Methyl 4-amino-1,2,5-thiadiazole-3-carboxylate

  • MDL number No MDL found
  • Cas Number : 63875-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40143.html

Safety Information

MSDS https://synthonix.com/40143.html
Availability details for
Quantity
  • Availability
  • Add to Cart