SY3H3D67F66B Synthonix Corporation

2-Methylthiazole

  • MDL number No MDL found
  • Cas Number : 3581-87-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/40165.html

Safety Information

MSDS https://synthonix.com/40165.html
Availability details for
Quantity
  • Availability
  • Add to Cart