SY3H3D67F674 Synthonix Corporation

1-Methyl-1H-pyrazol-5(4H)-one

  • MDL number No MDL found
  • Cas Number : 10234-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40183.html

Safety Information

MSDS https://synthonix.com/40183.html
Availability details for
Quantity
  • Availability
  • Add to Cart