SY3H3D67F67B Synthonix Corporation

(5-Methyl-1H-pyrazol-3-yl)boronic acid

  • MDL number No MDL found
  • Cas Number : 1163248-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40217.html

Safety Information

MSDS https://synthonix.com/40217.html
Availability details for
Quantity
  • Availability
  • Add to Cart