SY3H3D67F682 Synthonix Corporation

2-((Methoxycarbonyl)amino)acetic acid

  • Cas Number : 1670-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40229.html

Safety Information

MSDS https://synthonix.com/40229.html
Availability details for
Quantity
  • Availability
  • Add to Cart