SY3H3D67F683 Synthonix Corporation

Methyl 2-bromo-3-hydroxypropanoate

  • MDL number No MDL found
  • Cas Number : 7691-28-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40234.html

Safety Information

MSDS https://synthonix.com/40234.html
Availability details for
Quantity
  • Availability
  • Add to Cart