SY3H3D67F686 Synthonix Corporation

1-Methanesulfonyl-2-imidazolidinone

  • MDL number No MDL found
  • Cas Number : 41730-79-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40244.html

Safety Information

MSDS https://synthonix.com/40244.html
Availability details for
Quantity
  • Availability
  • Add to Cart