SY3H3D67F687 Synthonix Corporation

Methyl 2-(2-chloro-1-iminoethyl)hydrazinecarboxylate

  • MDL number No MDL found
  • Cas Number : 155742-64-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40245.html

Safety Information

MSDS https://synthonix.com/40245.html
Availability details for
Quantity
  • Availability
  • Add to Cart