SY3H3D67F68A Synthonix Corporation

(S)-Methyl 2,3-dihydroxypropanoate

  • MDL number No MDL found
  • Cas Number : 10303-88-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/40259.html

Safety Information

MSDS https://synthonix.com/40259.html
Availability details for
Quantity
  • Availability
  • Add to Cart