SY3H3D67F68E Synthonix Corporation

1-Methyl-4,5-dihydro-1H-imidazol-2-amine hydrobromide

  • MDL number No MDL found
  • Cas Number : 34122-57-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/40301.html

Safety Information

MSDS https://synthonix.com/40301.html
Availability details for
Quantity
  • Availability
  • Add to Cart