SY3H3D67F68F Synthonix Corporation

(R)-1-Methoxypropan-2-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 626220-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40311.html

Safety Information

MSDS https://synthonix.com/40311.html
Availability details for
Quantity
  • Availability
  • Add to Cart