SY3H3D67F690 Synthonix Corporation

(R)-2-Methoxypropan-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 907545-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40312.html

Safety Information

MSDS https://synthonix.com/40312.html
Availability details for
Quantity
  • Availability
  • Add to Cart