SY3H3D67F691 Synthonix Corporation

(S)-2-Methoxypropan-1-amine hydrochloride

  • MDL number No MDL found
  • Cas Number : 907544-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/40313.html

Safety Information

MSDS https://synthonix.com/40313.html
Availability details for
Quantity
  • Availability
  • Add to Cart