SY3H3D67F697 Synthonix Corporation

1-Methyl-1H-pyrazol-4-amine dihydrochloride

  • MDL number MFCD09025437
  • Cas Number : 1063734-49-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Light-yellow or light-red to brown powder or crystals
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/40360.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/40360.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart