SY3H3D67F69A Synthonix Corporation

(R)-2-Methylbutanoic acid

  • Cas Number : 32231-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40381.html

Safety Information

MSDS https://synthonix.com/40381.html
Availability details for
Quantity
  • Availability
  • Add to Cart