SY3H3D67F6A0 Synthonix Corporation

(S)-Methyl 2-aminobutanoate

  • MDL number No MDL found
  • Cas Number : 15399-22-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/40408.html

Safety Information

MSDS https://synthonix.com/40408.html
Availability details for
Quantity
  • Availability
  • Add to Cart