SY3H3D67F6A2 Synthonix Corporation

Methyl 3-(methylamino)propanoate

  • Cas Number : 24549-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40412.html

Safety Information

MSDS https://synthonix.com/40412.html
Availability details for
Quantity
  • Availability
  • Add to Cart