SY3H3D67F6A8 Synthonix Corporation

Methyl 2,5-dibromo-1H-imidazole-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 883876-21-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40756.html

Safety Information

MSDS https://synthonix.com/40756.html
Availability details for
Quantity
  • Availability
  • Add to Cart