SY3H3D67F6A9 Synthonix Corporation

5-Methyl-4-nitro-3-isoxazolecarboxylic acid

  • Cas Number : 960225-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/40768.html

Safety Information

MSDS https://synthonix.com/40768.html
Availability details for
Quantity
  • Availability
  • Add to Cart