SY3H3D67F6AA Synthonix Corporation

3-Methylisothiazole-4-carbonitrile

  • MDL number No MDL found
  • Cas Number : 58850-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/40783.html

Safety Information

MSDS https://synthonix.com/40783.html
Availability details for
Quantity
  • Availability
  • Add to Cart