SY3H3D67F6AC Synthonix Corporation

Methyl 5-chlorothiazole-4-carboxylate

  • MDL number No MDL found
  • Cas Number : 1784463-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/40839.html

Safety Information

MSDS https://synthonix.com/40839.html
Availability details for
Quantity
  • Availability
  • Add to Cart